Assay Details
| Gene Symbol: 9330158H04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015589.1 | 4 - 5 | 567 | 58 | ||
| GenBank mRNA | AK034128.1 | - | 4 - 5 | 568 | 58 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | myotrophin | Mtpn |
5033418D15Rik, Gcdp, V1 |
-796.4kb | Sense | Chr. 6 - 35508824 - 35539888 on NCBI Build GRCm38 |
| Downstream |
cholinergic receptor, muscarinic 2, cardiac |
Chrm2 |
AChR-M2, Chrm-2, M2 |
39.5kb | Anti-sense | Chr. 6 - 36387950 - 36528638 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

9330158H04Rik
Genomic Map