Assay Details
| Gene Symbol: Gls2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033264.3 | NP_001028436.2 | 1 - 2 | 274 | 118 | |
| NM_001285777.1 | NP_001272706.1 | 1 - 2 | 274 | 118 | ||
| NM_001285779.1 | NP_001272708.1 | 1 - 2 | 274 | 118 | ||
| GenBank mRNA | AK039618.1 | - | 1 - 2 | 162 | 118 | |
| AK165896.1 | - | 1 - 2 | 220 | 118 | ||
| AK220231.1 | - | 1 - 2 | 226 | 118 | ||
| BC141115.1 | - | 1 - 2 | 274 | 9055727 | 118 | |
| EU770627.1 | - | 1 - 2 | 183 | 118 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001285777.1, NM_001285779.1 transcripts.
Set Membership: |
|

Gls2
Genomic Map