Assay Details
| Gene Symbol: Gls2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033264.3 | NP_001028436.2 | 9 - 10 | 1017 | 68 | |
| NM_001285777.1 | NP_001272706.1 | 8 - 9 | 937 | 68 | ||
| NM_001285779.1 | NP_001272708.1 | 9 - 10 | 1046 | 68 | ||
| GenBank mRNA | AK039618.1 | - | 8 - 9 | 825 | 68 | |
| AK168761.1 | - | 2 - 3 | 149 | 68 | ||
| AK220231.1 | - | 9 - 10 | 969 | 68 | ||
| BC141115.1 | - | 9 - 10 | 1017 | 9055727 | 68 | |
| EU770627.1 | - | 9 - 10 | 955 | 68 | ||
More Information
Set Membership: |
|

Gls2
Genomic Map