Assay Details
| Gene Symbol: Cdk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007668.3 | NP_031694.1 | 5 - 6 | 350 | 77 | |
| GenBank mRNA | AK051790.1 | - | 5 - 6 | 350 | 77 | |
| BC052007.1 | - | 5 - 6 | 324 | 5706932 | 77 | |
| D29678.1 | - | 5 - 6 | 359 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdk5
Genomic Map