Assay Details
| Gene Symbol: Cpq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018755.2 | NP_061225.2 | 7 - 8 | 1354 | 73 | |
| NM_176073.4 | NP_788262.1 | 6 - 7 | 1264 | 73 | ||
| GenBank mRNA | AF009513.1 | - | 6 - 7 | 1109 | 73 | |
| AF107832.1 | - | 3 - 4 | 233 | 73 | ||
| AF107835.1 | - | 6 - 7 | 1232 | 73 | ||
| AK032972.1 | - | 6 - 7 | 1129 | 73 | ||
| AK075686.1 | - | 6 - 7 | 1195 | 73 | ||
| AK135096.1 | - | 6 - 7 | 1251 | 73 | ||
| AK137164.1 | - | 6 - 7 | 1138 | 73 | ||
| AK152139.1 | - | 7 - 8 | 1232 | 73 | ||
| AK152145.1 | - | 7 - 8 | 1232 | 73 | ||
| BC010977.1 | - | 6 - 7 | 1249 | 73 | ||
| BC037067.1 | - | 6 - 7 | 1111 | 4989536 | 73 | |
More Information
Set Membership: |
|

Cpq
Genomic Map