Assay Details
| Gene Symbol: Aass | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013930.4 | NP_038958.2 | 2 - 3 | 330 | 90 | |
| GenBank mRNA | AJ224761.1 | - | 1 - 2 | 221 | 90 | |
| AK044323.1 | - | 2 - 3 | 311 | 90 | ||
| AK098141.1 | - | 2 - 3 | 271 | 90 | ||
| AK136216.1 | - | 2 - 3 | 279 | 90 | ||
| AK149400.1 | - | 2 - 3 | 255 | 90 | ||
| BC005420.1 | - | 2 - 3 | 316 | 3600654 | 90 | |
| BY275733.1 | - | 2 - 3 | 318 | 90 | ||
| CJ074697.1 | - | 2 - 3 | 330 | 90 | ||
More Information
Set Membership: |
|

Aass
Genomic Map