Assay Details
| Gene Symbol: Lrig3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177152.5 | NP_796126.4 | 11 - 12 | 1574 | 67 | |
| GenBank mRNA | AK018522.1 | - | 5 - 6 | 387 | 67 | |
| AK129481.1 | - | 6 - 7 | 556 | 67 | ||
| AK137007.1 | - | 11 - 12 | 1560 | 67 | ||
| AY505341.1 | - | 11 - 12 | 1517 | 67 | ||
| BC065142.1 | - | 11 - 12 | 1540 | 30610561 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrig3
Genomic Map