Assay Details
| Gene Symbol: Lrig3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177152.5 | NP_796126.4 | 12 - 13 | 1734 | 79 | |
| GenBank mRNA | AK018522.1 | - | 6 - 7 | 547 | 79 | |
| AK129481.1 | - | 7 - 8 | 716 | 79 | ||
| AK137007.1 | - | 12 - 13 | 1720 | 79 | ||
| AY505341.1 | - | 12 - 13 | 1677 | 79 | ||
| BC065142.1 | - | 12 - 13 | 1700 | 30610561 | 79 | |
More Information
Set Membership: |
|

Lrig3
Genomic Map