Assay Details
| Gene Symbol: Lrig3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177152.5 | NP_796126.4 | 14 - 15 | 2341 | 73 | |
| GenBank mRNA | AK018522.1 | - | 8 - 9 | 1154 | 73 | |
| AK129481.1 | - | 10 - 11 | 1290 | 73 | ||
| AY505341.1 | - | 14 - 15 | 2284 | 73 | ||
| BC065142.1 | - | 14 - 15 | 2307 | 30610561 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrig3
Genomic Map