Assay Details
| Gene Symbol: Lrig3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177152.5 | NP_796126.4 | 15 - 16 | 2794 | 101 | |
| GenBank mRNA | AK018522.1 | - | 9 - 10 | 1607 | 101 | |
| AK078783.1 | - | 1 - 2 | 388 | 101 | ||
| AK129481.1 | - | 11 - 12 | 1743 | 101 | ||
| AY505341.1 | - | 15 - 16 | 2737 | 101 | ||
| BC065142.1 | - | 15 - 16 | 2760 | 30610561 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrig3
Genomic Map