Assay Details
| Gene Symbol: Lrig3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177152.5 | NP_796126.4 | 7 - 8 | 1198 | 100 | |
| GenBank mRNA | AK035166.1 | - | 7 - 8 | 1200 | 100 | |
| AK129481.1 | - | 2 - 3 | 182 | 100 | ||
| AK137007.1 | - | 7 - 8 | 1185 | 100 | ||
| AY505341.1 | - | 7 - 8 | 1141 | 100 | ||
| BC065142.1 | - | 7 - 8 | 1164 | 30610561 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrig3
Genomic Map