Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600016242.mm.1 |
TC0600002059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600016242.mm.1 |
NM_001164411.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600016242.mm.1 |
NM_001164411.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600016242.mm.1 |
NM_001164412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600016242.mm.1 |
NM_001164412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600016242.mm.1 |
NM_001164413.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600016242.mm.1 |
NM_001164413.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600016242.mm.1 |
NM_001164414.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600016242.mm.1 |
NM_001164414.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600016242.mm.1 |
NM_016877.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600016242.mm.1 |
NM_016877.4 |
Refseq |