Assay Details
| Gene Symbol: Tfpi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009364.3 | NP_033390.1 | 1 - 2 | 118 | 63 | |
| GenBank mRNA | AK132268.1 | - | 1 - 2 | 140 | 63 | |
| BC021639.1 | - | 1 - 2 | 153 | 4023742 | 63 | |
| D50586.1 | - | 1 - 2 | 123 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tfpi2
Genomic Map