Assay Details
| Gene Symbol: Rpa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026632.4 | NP_080908.1 | 3 - 4 | 324 | 119 | |
| GenBank mRNA | AK009916.1 | - | 3 - 4 | 296 | 119 | |
| AK021228.1 | - | 3 - 4 | 297 | 119 | ||
| BC028489.1 | - | 3 - 4 | 289 | 1448430 | 119 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair via homologous recombination
DNA replication
DNA repair
base-excision repair
nucleotide-excision repair
mismatch repair
DNA recombination
cellular response to DNA damage stimulus
regulation of mitotic cell cycle
regulation of cell proliferation
DNA binding
damaged DNA binding
single-stranded DNA binding
double-strand break repair via homologous recombination
DNA replication
DNA repair
base-excision repair
nucleotide-excision repair
mismatch repair
DNA recombination
cellular response to DNA damage stimulus
regulation of mitotic cell cycle
regulation of cell proliferation
DNA binding
damaged DNA binding
single-stranded DNA binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24225084 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24225085 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24225082 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24225086 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P106090 | BC028489.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P106090 | AK021228.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P106090 | BY057686.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P106090 | NM_026632.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P106090 | AK005285.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 815403 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 912682 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 432957 | AF373861.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 432957 | AF396649.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 432957 | AK131170.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 432957 | AY062435.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 432957 | BC115747.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 432957 | NM_130455.2 | refseq_mrna_mouse_NM |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135488 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P106090 | BC028489.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P106090 | AK021228.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P106090 | BY057686.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P106090 | NM_026632.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P106090 | AK005285.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P438057 | BC028489.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P438057 | AK021228.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P438057 | NM_026632.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P438057 | AK005285.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P438057 | AK009916.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24225081 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24225055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24225061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24225062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24225072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24225066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24225063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24225073 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24225082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24225057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24225056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24225086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24225080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24225078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24225067 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24225053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24225070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24225064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24225085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24225068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24225076 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135488 | - | - |
| Whole Mouse Genome G4122A | A_51_P438057 | BC028489.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P438057 | AK021228.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P438057 | NM_026632.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P438057 | AK005285.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P438057 | AK009916.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106090 | BC028489.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106090 | AK021228.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106090 | BY057686.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106090 | NM_026632.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106090 | AK005285.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135488 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05665498 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05665507 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05665502 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05665499 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24225085 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24225082 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24225086 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24225084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24225110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225106 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225094 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225134 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225064 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225060 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225031 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225128 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225049 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05266678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225051 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05665502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05665508 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225063 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225115 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225095 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05173242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225077 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225117 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225100 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225035 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225048 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05359289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225119 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225099 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225118 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225068 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05324486 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225131 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05665491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05025376 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225121 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05173243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225112 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225062 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225040 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225085 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05507455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225056 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05771502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225108 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225057 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225107 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225126 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225103 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225098 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24225041 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10543116 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10543115 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353033 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10543117 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448938_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10543114 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10353032 | - | - |
| Mouse Genome 430 2.0 Array | 1448938_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448938_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68240_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68240_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600014559.mm.1 | TC0600001844.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600014559.mm.1 | NM_026632.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600014559.mm.1 | NM_026632.4 | Refseq |
| Mouse Expression Set 430 | 1448938_at | - | - |
| Mouse Expression Set 430 | 1448938_PM_at | - | - |
| Murine Genome U74v2 Set | 160324_PM_at | - | - |
| Murine Genome U74v2 Set | 160324_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448938_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448938_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448938_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448938_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5700136 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5700136 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005700136 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1239806 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1239806 | - | - |
| mouse-6 v1.1 (Illumina) | scl068240.2_29-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5700136 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl068240.2_29-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5700136 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1239806 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1239806 | - | - |

Rpa3
Genomic Map