Assay Details
| Gene Symbol: Cd36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159555.1 | NP_001153027.1 | 1 - 2 | 272 | 137 | |
| NM_001159557.1 | NP_001153029.1 | 2 - 3 | 183 | 137 | ||
| GenBank mRNA | AK151355.1 | - | 1 - 2 | 111 | 137 | |
| AK172162.1 | - | 2 - 3 | 121 | 137 | ||
| AW986994.1 | - | 1 - 2 | 272 | 137 | ||
| L23108.1 | - | 1 - 2 | 93 | 137 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK151355.1, AK172162.1, L23108.1, NM_001159555.1, NM_001159557.1 transcripts.
Set Membership: |
|

Cd36
Genomic Map