Assay Details
Gene Symbol: Hyal5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028957.2 | NP_083233.1 | 1 - 2 | 114 | 129 | |
GenBank mRNA | AK017112.1 | - | 1 - 2 | 129 | 129 | |
AK133387.1 | - | 1 - 2 | 117 | 129 | ||
BC145992.1 | - | 1 - 2 | 47 | 40131295 | 129 | |
BC145994.1 | - | 1 - 2 | 47 | 40131297 | 129 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK017112.1, AK133387.1, BC145992.1, BC145994.1, NM_028957.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
