Assay Details
| Gene Symbol: Hyal6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028920.2 | NP_083196.2 | 1 - 2 | 274 | 91 | |
| GenBank mRNA | AK016575.1 | - | 1 - 2 | 275 | 91 | |
| AK029715.1 | - | 1 - 2 | 225 | 91 | ||
| AK030190.1 | - | 1 - 2 | 275 | 91 | ||
| BC151178.1 | - | 1 - 2 | 212 | 9088067 | 91 | |
| BC151179.1 | - | 1 - 2 | 212 | 9088068 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK016575.1, AK029715.1, BC151178.1, BC151179.1, NM_028920.2 transcripts.
Set Membership: |
|

Hyal6
Genomic Map