Assay Details
| Gene Symbol: Hyal6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028920.2 | NP_083196.2 | 3 - 4 | 1323 | 90 | |
| GenBank mRNA | AK016575.1 | - | 3 - 4 | 1324 | 90 | |
| AK029715.1 | - | 3 - 4 | 1274 | 90 | ||
| AK030190.1 | - | 3 - 4 | 1324 | 90 | ||
| BC151178.1 | - | 3 - 4 | 1261 | 9088067 | 90 | |
| BC151179.1 | - | 3 - 4 | 1261 | 9088068 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2052325 | AK029715.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2052325 | NM_028920.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 707254 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 484007 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 69148 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 504633 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 300517 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 229259 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 139855 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 28783 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100990 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171954 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2052325 | AK029715.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2052325 | NM_028920.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303271 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24303274 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05837494 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171954 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100990 | - | - |
| Whole Mouse Genome G4122A | A_51_P505986 | BC151178.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505986 | BC151179.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505986 | AK029715.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505986 | AK030190.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505986 | NM_028920.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171954 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100990 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05135408 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05247068 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05247047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303314 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05024439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303325 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05350736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05247052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05832386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05116527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05882034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303248 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05035835 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05350737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303317 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10536719 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536720 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536718 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536721 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430049_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10536717 | - | - |
| Mouse Genome 430 2.0 Array | 1430049_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430049_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74409_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74409_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600001291.mm.1 | TC0600000203.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600001291.mm.1 | NM_028920.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600001291.mm.1 | NM_028920.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600001292.mm.1 | TC0600000203.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600001292.mm.1 | NM_028920.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600001292.mm.1 | NM_028920.2 | Refseq |
| Mouse Expression Set 430 | 1430049_at | - | - |
| Mouse Expression Set 430 | 1430049_PM_at | - | - |
| Murine Genome U74v2 Set | 166359_PM_at | - | - |
| Murine Genome U74v2 Set | 166359_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430049_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430049_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 750955 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 105220097 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105220097 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101580161 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1234019 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1242948 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1227971 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101580161 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105220097 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101580161 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105220097 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2966519 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1234019 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1227971 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2966519 | - | - |

Hyal6
Genomic Map