Assay Details
| Gene Symbol: Cdh12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008420.2 | NP_001008420.1 | 6 - 7 | 1198 | 101 | |
| GenBank mRNA | BC086655.1 | - | 6 - 7 | 1198 | 30542783 | 101 |
More Information
Updated Mapping Information
Set Membership: |
|

Cdh12
Genomic Map