Assay Details
| Gene Symbol: Dchs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162943.1 | NP_001156415.1 | 11 - 12 | 5324 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dchs1| Gene Symbol: Dchs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162943.1 | NP_001156415.1 | 11 - 12 | 5324 | 94 | |
Set Membership: |
|