Assay Details
| Gene Symbol: Rabep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030566.2 | NP_085043.2 | 6 - 7 | 1308 | 79 | |
| GenBank mRNA | AK029188.1 | - | 4 - 5 | 666 | 79 | |
| AK089938.1 | - | 6 - 7 | 1012 | 79 | ||
| AK144836.1 | - | 6 - 7 | 994 | 79 | ||
| AK157832.1 | - | 6 - 7 | 969 | 79 | ||
| AK218661.1 | - | 1 - 2 | 48 | 79 | ||
| BC004088.1 | - | 5 - 6 | 1054 | 3593722 | 79 | |
| BC015287.1 | - | 6 - 7 | 1316 | 4240442 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rabep2
Genomic Map