Assay Details
| Gene Symbol: Rabep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030566.2 | NP_085043.2 | 10 - 11 | 1761 | 67 | |
| GenBank mRNA | AK089938.1 | - | 10 - 11 | 1466 | 67 | |
| AK144836.1 | - | 10 - 11 | 1447 | 67 | ||
| AK157832.1 | - | 10 - 11 | 1422 | 67 | ||
| BC004088.1 | - | 9 - 10 | 1507 | 3593722 | 67 | |
| BC015287.1 | - | 10 - 11 | 1769 | 4240442 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rabep2
Genomic Map