| Array Name |
Probe IDs |
Targets |
Target Type |
| Mouse Genome 430 2.0 Array |
1430067_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1430067_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0600012580.mm.1 |
TC0600001551.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600012580.mm.1 |
NM_001244628.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012580.mm.1 |
NM_001244628.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012583.mm.1 |
TC0600001551.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600012583.mm.1 |
NM_001244628.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012583.mm.1 |
NM_001244628.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012581.mm.1 |
TC0600001551.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600012581.mm.1 |
NM_001244628.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012581.mm.1 |
NM_001244628.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012582.mm.1 |
TC0600001551.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600012582.mm.1 |
NM_001244628.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012582.mm.1 |
NM_001244628.1 |
Refseq |
| Mouse Expression Set 430 |
1430067_at |
- |
- |
| Mouse Expression Set 430 |
1430067_PM_at |
- |
- |
| Murine Genome U74v2 Set |
166609_at |
- |
- |
| Murine Genome U74v2 Set |
166609_PM_at |
- |
- |