Assay Details
| Gene Symbol: Trim68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK164544.1 | - | 2 - 3 | 600 | 75 | |
| BC034249.1 | - | 2 - 3 | 583 | 3675490 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim68
Genomic Map