Assay Details
| Gene Symbol: Trim68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC034249.1 | - | 4 - 5 | 933 | 3675490 | 79 |
| CO424835.1 | - | 2 - 3 | 412 | 79 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim68| Gene Symbol: Trim68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC034249.1 | - | 4 - 5 | 933 | 3675490 | 79 |
| CO424835.1 | - | 2 - 3 | 412 | 79 | ||
Set Membership: |
|