Assay Details
| Gene Symbol: Mrs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013389.2 | NP_001013407.2 | 6 - 7 | 769 | 93 | |
| GenBank mRNA | AK136100.1 | - | 1 - 2 | 60 | 93 | |
| CA752560.1 | - | 3 - 4 | 422 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mrs2
Genomic Map