Assay Details
| Gene Symbol: Arhgef5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133674.1 | NP_598435.1 | 12 - 13 | 4525 | 71 | |
| GenBank mRNA | AK037202.1 | - | 7 - 8 | 942 | 71 | |
| AK220242.1 | - | 1 - 2 | 1442 | 71 | ||
| BC025127.1 | - | 7 - 8 | 812 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef5
Genomic Map