Assay Details
| Gene Symbol: Arhgef5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133674.1 | NP_598435.1 | 13 - 14 | 4600 | 89 | |
| GenBank mRNA | AK020295.1 | - | 1 - 2 | 364 | 89 | |
| AK220242.1 | - | 2 - 3 | 1517 | 89 | ||
| BC006829.1 | - | 7 - 8 | 2295 | 89 | ||
| BC025127.1 | - | 8 - 9 | 887 | 89 | ||
| CB058042.1 | - | 1 - 2 | 78 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef5
Genomic Map