Assay Details
| Gene Symbol: Arhgef5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133674.1 | NP_598435.1 | 5 - 6 | 3598 | 84 | |
| GenBank mRNA | AK008907.1 | - | 1 - 2 | 127 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef5
Genomic Map