Assay Details
| Gene Symbol: Arhgef5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133674.1 | NP_598435.1 | 8 - 9 | 4036 | 60 | |
| GenBank mRNA | AK037202.1 | - | 3 - 4 | 453 | 60 | |
| BC006829.1 | - | 3 - 4 | 427 | 60 | ||
| BC025127.1 | - | 3 - 4 | 323 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef5
Genomic Map