Assay Details
| Gene Symbol: Chid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142681.1 | NP_001136153.1 | 10 - 11 | 1082 | 73 | |
| GenBank mRNA | AK036717.1 | - | 10 - 11 | 1027 | 73 | |
| AK147377.1 | - | 8 - 9 | 5880 | 73 | ||
| AK160156.1 | - | 10 - 11 | 1042 | 73 | ||
| AK160200.1 | - | 10 - 11 | 1043 | 73 | ||
| BC006876.1 | - | 10 - 11 | 1031 | 2647540 | 73 | |
| BC061063.1 | - | 10 - 11 | 1042 | 6591338 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chid1
Genomic Map