Assay Details
| Gene Symbol: Ctnnd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008729.2 | NP_032755.2 | 18 - 19 | 3597 | 59 | |
| GenBank mRNA | AK140454.1 | - | 8 - 9 | 1215 | 59 | |
| AK140960.1 | - | 3 - 4 | 472 | 59 | ||
| AK164304.1 | - | 1 - 2 | 148 | 59 | ||
| BC138260.1 | - | 18 - 19 | 3142 | 8861279 | 59 | |
| BC138261.1 | - | 18 - 19 | 3142 | 8861280 | 59 | |
| BC145217.1 | - | 17 - 18 | 3064 | 9053768 | 59 | |
| U90331.1 | - | 18 - 19 | 3600 | 59 | ||
More Information
Set Membership: |
|

Ctnnd2
Genomic Map