Assay Details
| Gene Symbol: Ctnnd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008729.2 | NP_032755.2 | 9 - 10 | 2149 | 59 | |
| GenBank mRNA | BC138260.1 | - | 9 - 10 | 1694 | 8861279 | 59 |
| BC138261.1 | - | 9 - 10 | 1694 | 8861280 | 59 | |
| BC145217.1 | - | 9 - 10 | 1691 | 9053768 | 59 | |
| U90331.1 | - | 9 - 10 | 2152 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctnnd2
Genomic Map