Assay Details
| Gene Symbol: Copg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017478.3 | NP_059506.1 | 1745 | 70 | ||
| GenBank mRNA | AF205065.1 | - | 1648 | 70 | ||
| AJ251067.1 | - | 1717 | 70 | |||
| AK088492.1 | - | 1729 | 70 | |||
| AK147943.1 | - | 1692 | 70 | |||
| BC049178.1 | - | 1711 | 6309945 | 70 | ||
| BC145898.1 | - | 1688 | 40131201 | 70 | ||
| BC145900.1 | - | 1688 | 40131203 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Copg2
Genomic Map