Assay Details
| Gene Symbol: Copg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017478.3 | NP_059506.1 | 2073 | 80 | ||
| GenBank mRNA | AF205065.1 | - | 1976 | 80 | ||
| AJ251067.1 | - | 2045 | 80 | |||
| AK088492.1 | - | 2057 | 80 | |||
| AK147943.1 | - | 2020 | 80 | |||
| BC049178.1 | - | 2039 | 6309945 | 80 | ||
| BC145898.1 | - | 2016 | 40131201 | 80 | ||
| BC145900.1 | - | 2016 | 40131203 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Copg2
Genomic Map