Assay Details
| Gene Symbol: Slc37a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC005744.1 | - | 6 - 7 | 499 | 61 | |
| BC100391.1 | - | 7 - 8 | 714 | 61 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc37a3| Gene Symbol: Slc37a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC005744.1 | - | 6 - 7 | 499 | 61 | |
| BC100391.1 | - | 7 - 8 | 714 | 61 | ||
Set Membership: |
|