Assay Details
| Gene Symbol: Pip4k2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054097.3 | NP_473438.1 | 2 - 3 | 384 | 116 | |
| GenBank mRNA | AB054591.1 | - | 2 - 3 | 327 | 116 | |
| AK140079.1 | - | 2 - 3 | 348 | 116 | ||
| BC021383.1 | - | 2 - 3 | 317 | 5042933 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pip4k2c
Genomic Map