Assay Details
| Gene Symbol: Pip4k2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054097.3 | NP_473438.1 | 3 - 4 | 478 | 80 | |
| GenBank mRNA | AB054591.1 | - | 3 - 4 | 421 | 80 | |
| AK148299.1 | - | 3 - 4 | 438 | 80 | ||
| AK207886.1 | - | 1 - 2 | 86 | 80 | ||
| BC021383.1 | - | 3 - 4 | 411 | 5042933 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pip4k2c
Genomic Map