Assay Details
| Gene Symbol: Pip4k2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054097.3 | NP_473438.1 | 6 - 7 | 816 | 127 | |
| GenBank mRNA | AB054591.1 | - | 6 - 7 | 759 | 127 | |
| AK148299.1 | - | 6 - 7 | 776 | 127 | ||
| AK208052.1 | - | 3 - 4 | 229 | 127 | ||
| BC005539.1 | - | 1 - 2 | 30 | 127 | ||
More Information
Set Membership: |
|

Pip4k2c
Genomic Map