Assay Details
| Gene Symbol: Mars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003913.2 | NP_001003913.1 | 10 - 11 | 1457 | 62 | |
| NM_001171582.1 | NP_001165053.1 | 10 - 11 | 1457 | 62 | ||
| GenBank mRNA | AK192160.1 | - | 1 - 2 | 115 | 62 | |
| BC052042.1 | - | 10 - 11 | 1309 | 62 | ||
| BC058968.1 | - | 10 - 11 | 1349 | 62 | ||
| BC079643.1 | - | 10 - 11 | 1310 | 6416029 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mars
Genomic Map