Assay Details
| Gene Symbol: Mars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003913.2 | NP_001003913.1 | 11 - 12 | 1537 | 120 | |
| NM_001171582.1 | NP_001165053.1 | 11 - 12 | 1537 | 120 | ||
| GenBank mRNA | AK192160.1 | - | 2 - 3 | 195 | 120 | |
| BC052042.1 | - | 11 - 12 | 1389 | 120 | ||
| BC058968.1 | - | 11 - 12 | 1429 | 120 | ||
| BC079643.1 | - | 11 - 12 | 1390 | 6416029 | 120 | |
More Information
Set Membership: |
|

Mars
Genomic Map