Assay Details
| Gene Symbol: Mars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003913.2 | NP_001003913.1 | 12 - 13 | 1702 | 67 | |
| NM_001171582.1 | NP_001165053.1 | 12 - 13 | 1702 | 67 | ||
| GenBank mRNA | BC052042.1 | - | 12 - 13 | 1554 | 67 | |
| BC058968.1 | - | 12 - 13 | 1594 | 67 | ||
| BC079643.1 | - | 12 - 13 | 1555 | 6416029 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mars
Genomic Map