Assay Details
| Gene Symbol: Mars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003913.2 | NP_001003913.1 | 14 - 15 | 1914 | 74 | |
| NM_001171582.1 | NP_001165053.1 | 14 - 15 | 1914 | 74 | ||
| GenBank mRNA | AK191707.1 | - | 2 - 3 | 177 | 74 | |
| AK197362.1 | - | 2 - 3 | 178 | 74 | ||
| BC052042.1 | - | 14 - 15 | 1766 | 74 | ||
| BC058968.1 | - | 14 - 15 | 1806 | 74 | ||
| BC079643.1 | - | 14 - 15 | 1767 | 6416029 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mars
Genomic Map