Assay Details
| Gene Symbol: Mars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171582.1 | NP_001165053.1 | 20 - 21 | 2652 | 84 | |
| GenBank mRNA | AA510607.1 | - | 3 - 4 | 218 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mars
Genomic Map