Assay Details
| Gene Symbol: Mars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003913.2 | NP_001003913.1 | 3 - 4 | 442 | 72 | |
| NM_001171582.1 | NP_001165053.1 | 3 - 4 | 442 | 72 | ||
| GenBank mRNA | AK181384.1 | - | 2 - 3 | 152 | 72 | |
| AK213658.1 | - | 2 - 3 | 153 | 72 | ||
| BC052042.1 | - | 3 - 4 | 294 | 72 | ||
| BC058968.1 | - | 3 - 4 | 334 | 72 | ||
| BC079643.1 | - | 3 - 4 | 295 | 6416029 | 72 | |
More Information
Set Membership: |
|

Mars
Genomic Map