Assay Details
| Gene Symbol: Mars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003913.2 | NP_001003913.1 | 5 - 6 | 646 | 68 | |
| NM_001171582.1 | NP_001165053.1 | 5 - 6 | 646 | 68 | ||
| GenBank mRNA | AK178137.1 | - | 3 - 4 | 286 | 68 | |
| AK184017.1 | - | 2 - 3 | 128 | 68 | ||
| AK189447.1 | - | 4 - 5 | 357 | 68 | ||
| AK193348.1 | - | 2 - 3 | 158 | 68 | ||
| AK214150.1 | - | 3 - 4 | 243 | 68 | ||
| BC052042.1 | - | 5 - 6 | 498 | 68 | ||
| BC058968.1 | - | 5 - 6 | 538 | 68 | ||
| BC079643.1 | - | 5 - 6 | 499 | 6416029 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mars
Genomic Map