Assay Details
| Gene Symbol: March9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033262.2 | NP_001028434.1 | 1 - 2 | 793 | 74 | |
| GenBank mRNA | AK158026.1 | - | 1 - 2 | 796 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

March9
Genomic Map