Assay Details
| Gene Symbol: March9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033262.2 | NP_001028434.1 | 3 - 4 | 1130 | 83 | |
| GenBank mRNA | AK158026.1 | - | 3 - 4 | 1133 | 83 | |
| BC019560.1 | - | 2 - 3 | 297 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
molecular_function
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity
biological_process
molecular_function
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533785 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533787 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533789 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533782 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533783 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533788 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2011922 | BC019560.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2011922 | NM_001033262.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2011922 | AK158026.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 419121 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 38272 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178354 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2011922 | BC019560.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2011922 | NM_001033262.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2011922 | AK158026.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533801 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533796 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533808 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533793 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533791 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533799 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533806 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533787 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533776 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178354 | - | - |
| Whole Mouse Genome G4122A | A_51_P226794 | BC019560.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P226794 | NM_001033262.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P226794 | AK158026.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178354 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08846728 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08846725 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08846730 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08846735 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08846734 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533788 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533785 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533789 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533782 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533787 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08545505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08545500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08261412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08210286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533750 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08545511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08210289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08210284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08261410 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08261436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08210303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08261423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08210298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08545498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08261440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533798 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10373022 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373025 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373026 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373023 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437366_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427409_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10373021 | - | - |
| Mouse Genome 430 2.0 Array | 1427409_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1427409_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000023038.mm.1 | NM_001033262.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023038.mm.1 | NM_001033262.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023038.mm.1 | TC1000003095.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427409_PM_at | - | - |
| Mouse Expression Set 430 | 1427409_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427409_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427409_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2600900 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2600900 | - | - |

March9
Genomic Map