Assay Details
| Gene Symbol: Cyp27b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010009.2 | NP_034139.2 | 2 - 3 | 419 | 98 | |
| GenBank mRNA | AB006034.1 | - | 2 - 3 | 413 | 98 | |
| BC080697.1 | - | 2 - 3 | 454 | 30470456 | 98 | |
| CF168641.1 | - | 2 - 3 | 419 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyp27b1
Genomic Map