Assay Details
| Gene Symbol: Cyp27b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010009.2 | NP_034139.2 | 5 - 6 | 1002 | 81 | |
| GenBank mRNA | AB006034.1 | - | 5 - 6 | 996 | 81 | |
| BC080697.1 | - | 5 - 6 | 1037 | 30470456 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyp27b1
Genomic Map